Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077226.1 | NP_001070694.1 | 5 - 6 | 563 | 96 | |
XM_005664070.3 | XP_005664127.1 | 768 | 96 | |||
XM_013997827.2 | XP_013853281.1 | 1727 | 96 | |||
GenBank mRNA | AK348596.1 | - | 7 - 8 | 725 | 96 | |
AK396389.1 | - | 7 - 8 | 722 | 96 | ||
DQ972959.1 | - | 5 - 6 | 563 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |