Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078681.1 | NP_001072149.1 | 4 - 5 | 565 | 102 | |
XM_013982998.2 | XP_013838452.1 | 551 | 102 | |||
GenBank mRNA | DQ915497.1 | - | 4 - 5 | 565 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |