Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123136.1 | NP_001116608.1 | 4 - 5 | 393 | 140 | |
GenBank mRNA | AK233908.1 | - | 4 - 5 | 393 | 140 | |
AK346706.1 | - | 4 - 5 | 396 | 140 | ||
AK346872.1 | - | 4 - 5 | 373 | 140 | ||
AK347071.1 | - | 4 - 5 | 401 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |