Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213983.1 | NP_999148.1 | 8 - 9 | 1015 | 77 | |
XM_005668681.3 | XP_005668738.1 | 1098 | 77 | |||
XM_013980839.2 | XP_013836293.1 | 1147 | 77 | |||
XM_021066208.1 | XP_020921867.1 | 1100 | 77 | |||
XM_021066209.1 | XP_020921868.1 | 1151 | 77 | |||
XM_021066210.1 | XP_020921869.1 | 964 | 77 | |||
XM_021066211.1 | XP_020921870.1 | 975 | 77 | |||
GenBank mRNA | AK234194.1 | - | 8 - 9 | 905 | 77 | |
AK395258.1 | - | 8 - 9 | 927 | 77 | ||
AK396074.1 | - | 8 - 9 | 900 | 77 | ||
X89719.1 | - | 843 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |