Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXFP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005844.1 | NP_001005844.1 | 1 | 843 | 79 | |
GenBank mRNA | AK393003.1 | - | 4 | 565 | 79 | |
AY633768.1 | - | 1 | 843 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |