Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214066.2 | NP_999231.1 | 7 - 8 | 827 | 150 | |
GenBank mRNA | AK233002.1 | - | 7 - 8 | 700 | 150 | |
AK390956.1 | - | 7 - 8 | 809 | 150 | ||
AK391151.1 | - | 7 - 8 | 700 | 150 | ||
M16972.1 | - | 7 - 8 | 827 | 150 | ||
M18448.1 | - | 2 - 3 | 160 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |