Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214091.2 | NP_999256.1 | 1 - 2 | 75 | 75 | |
GenBank mRNA | AF484233.1 | - | 1 - 2 | 76 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |