Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214346.1 | NP_999511.1 | 8 - 9 | 2123 | 57 | |
XM_021077303.1 | XP_020932962.1 | 2387 | 57 | |||
XM_021077304.1 | XP_020932963.1 | 2389 | 57 | |||
GenBank mRNA | AF509585.1 | - | 8 - 9 | 2123 | 57 | |
DQ176853.1 | - | 8 - 9 | 2123 | 57 | ||
EU003993.1 | - | 8 - 9 | 2123 | 57 | ||
JX070181.1 | - | 8 - 9 | 2123 | 57 | ||
JX070182.1 | - | 8 - 9 | 2123 | 57 | ||
JX070183.1 | - | 8 - 9 | 2123 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |