Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR0B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214387.1 | NP_999552.1 | 1 - 2 | 1191 | 80 | |
GenBank mRNA | AF035816.1 | - | 2 - 3 | 1170 | 80 | |
U82466.1 | - | 1 - 2 | 1191 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Sus scrofa Probe spans exons |