Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166313.1 | NP_001159785.1 | 3 - 4 | 307 | 106 | |
XM_005661543.3 | XP_005661600.1 | 869 | 106 | |||
XM_005661544.3 | XP_005661601.1 | 536 | 106 | |||
XM_013994968.2 | XP_013850422.1 | 869 | 106 | |||
XM_013994969.2 | XP_013850423.1 | 869 | 106 | |||
XM_013994970.2 | XP_013850424.1 | 869 | 106 | |||
XM_013994971.2 | XP_013850425.1 | 869 | 106 | |||
XM_013994972.2 | XP_013850426.1 | 869 | 106 | |||
XM_021077281.1 | XP_020932940.1 | 869 | 106 | |||
XM_021077311.1 | XP_020932970.1 | 869 | 106 | |||
GenBank mRNA | AB499892.1 | - | 3 - 4 | 307 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |