Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146300.2 | NP_001139772.1 | 2 - 3 | 1109 | 96 | |
GenBank mRNA | AK236699.1 | - | 2 - 3 | 1111 | 96 | |
AK392031.1 | - | 2 - 3 | 1107 | 96 | ||
FJ790418.1 | - | 2 - 3 | 1111 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |