Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159615.1 | NP_001153087.1 | 5 - 6 | 938 | 60 | |
GenBank mRNA | AK350973.1 | - | 5 - 6 | 943 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |