Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185135.1 | NP_001172064.1 | 1 - 2 | 105 | 101 | |
GenBank mRNA | AK230934.1 | - | 2 - 3 | 159 | 101 | |
EU868836.1 | - | 1 - 2 | 105 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |