Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190235.1 | NP_001177164.1 | 3 - 4 | 431 | 75 | |
GenBank mRNA | AK236439.1 | - | 3 - 4 | 434 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |