Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204382.1 | NP_001191311.1 | 5 - 6 | 497 | 98 | |
XM_021090906.1 | XP_020946565.1 | 731 | 98 | |||
XM_021090908.1 | XP_020946567.1 | 738 | 98 | |||
XM_021090909.1 | XP_020946568.1 | 720 | 98 | |||
XM_021090910.1 | XP_020946569.1 | 461 | 98 | |||
XM_021090911.1 | XP_020946570.1 | 676 | 98 | |||
XM_021090912.1 | XP_020946571.1 | 729 | 98 | |||
XM_021090913.1 | XP_020946572.1 | 461 | 98 | |||
GenBank mRNA | AK239479.1 | - | 6 - 7 | 444 | 98 | |
AK350790.1 | - | 5 - 6 | 497 | 98 | ||
AK391116.1 | - | 5 - 6 | 653 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |