Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YKT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244914.1 | NP_001231843.1 | 6 - 7 | 1052 | 79 | |
GenBank mRNA | AK233852.1 | - | 6 - 7 | 1052 | 79 | |
AY609946.1 | - | 6 - 7 | 614 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |