Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIH1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244978.1 | NP_001231907.1 | 2 - 3 | 475 | 73 | |
XM_005664715.3 | XP_005664772.1 | 303 | 73 | |||
XM_005664716.3 | XP_005664773.1 | 300 | 73 | |||
GenBank mRNA | AK234303.1 | - | 2 - 3 | 475 | 73 | |
AK236765.1 | - | 2 - 3 | 493 | 73 | ||
AK348445.1 | - | 2 - 3 | 475 | 73 | ||
AY609770.1 | - | 6 - 7 | 724 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |