Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243407.1 | NP_001230336.1 | 5 - 6 | 663 | 79 | |
GenBank mRNA | AK349470.1 | - | 5 - 6 | 663 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |