Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243470.1 | NP_001230399.1 | 6 - 7 | 653 | 94 | |
GenBank mRNA | AK235362.1 | - | 5 - 6 | 607 | 94 | |
AK346494.1 | - | 6 - 7 | 653 | 94 | ||
AK394233.1 | - | 5 - 6 | 653 | 94 | ||
AK394808.1 | - | 5 - 6 | 676 | 94 | ||
AY610211.1 | - | 520 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |