Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYKK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243547.1 | NP_001230476.1 | 1 - 2 | 72 | 85 | |
GenBank mRNA | AK351146.1 | - | 1 - 2 | 72 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |