Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDARADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243663.1 | NP_001230592.1 | 2 - 3 | 209 | 74 | |
XM_021072012.1 | XP_020927671.1 | 1609 | 74 | |||
XM_021072013.1 | XP_020927672.1 | 539 | 74 | |||
GenBank mRNA | AK239039.1 | - | 2 - 3 | 209 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |