Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243711.1 | NP_001230640.1 | 1 - 2 | 87 | 88 | |
GenBank mRNA | AK396456.1 | - | 1 - 2 | 81 | 88 | |
FS721678.1 | - | 1 - 2 | 87 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |