Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM59L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243817.1 | NP_001230746.1 | 6 - 7 | 872 | 92 | |
XM_021077858.1 | XP_020933517.1 | 878 | 92 | |||
GenBank mRNA | AK349298.1 | - | 6 - 7 | 872 | 92 | |
AK392001.1 | - | 6 - 7 | 864 | 92 | ||
AK399981.1 | - | 6 - 7 | 887 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |