Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244107.1 | NP_001231036.1 | 5 - 6 | 685 | 104 | |
NM_001244109.1 | NP_001231038.1 | 3 - 4 | 500 | 104 | ||
XM_005673457.3 | XP_005673514.1 | 633 | 104 | |||
GenBank mRNA | AK233306.1 | - | 4 - 5 | 355 | 104 | |
AK235288.1 | - | 3 - 4 | 504 | 104 | ||
AK237134.1 | - | 4 - 5 | 355 | 104 | ||
AK240229.1 | - | 5 - 6 | 685 | 104 | ||
AK397391.1 | - | 5 - 6 | 685 | 104 | ||
AK399243.1 | - | 5 - 6 | 697 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |