Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244358.1 | NP_001231287.1 | 3 - 4 | 312 | 93 | |
GenBank mRNA | AK230977.1 | - | 3 - 4 | 416 | 93 | |
AK237144.1 | - | 3 - 4 | 331 | 93 | ||
AK238754.1 | - | 3 - 4 | 331 | 93 | ||
AK347683.1 | - | 3 - 4 | 312 | 93 | ||
AK399200.1 | - | 3 - 4 | 346 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |