Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244514.1 | NP_001231443.1 | 4 - 5 | 529 | 81 | |
GenBank mRNA | AK346775.1 | - | 4 - 5 | 507 | 81 | |
CJ011354.1 | - | 4 - 5 | 529 | 81 | ||
CV875205.1 | - | 1 - 2 | 73 | 81 | ||
FS702503.1 | - | 4 - 5 | 509 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |