Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244718.1 | NP_001231647.1 | 8 - 9 | 751 | 102 | |
NM_001244719.1 | NP_001231648.1 | 7 - 8 | 712 | 102 | ||
XM_005659700.3 | XP_005659757.1 | 887 | 102 | |||
XM_013993108.2 | XP_013848562.1 | 762 | 102 | |||
XM_013993109.2 | XP_013848563.1 | 948 | 102 | |||
XM_013993110.2 | XP_013848564.1 | 781 | 102 | |||
XM_013993111.2 | XP_013848565.1 | 941 | 102 | |||
XM_013993112.2 | XP_013848566.1 | 790 | 102 | |||
XM_021069852.1 | XP_020925511.1 | 878 | 102 | |||
XM_021070075.1 | XP_020925734.1 | 748 | 102 | |||
XM_021070141.1 | XP_020925800.1 | 621 | 102 | |||
GenBank mRNA | AK230671.1 | - | 8 - 9 | 751 | 102 | |
AK230848.1 | - | 7 - 8 | 692 | 102 | ||
AK389714.1 | - | 7 - 8 | 698 | 102 | ||
AY609458.1 | - | 7 - 8 | 712 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |