Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ490336.1 | - | 9 - 10 | 1684 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Vitis vinifera Probe spans exons |