Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ865335.1 | - | 2 - 3 | 336 | 72 | |
AJ865336.1 | - | 2 - 3 | 424 | 72 | ||
DQ129685.1 | - | 2 - 3 | 336 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Probe spans exons |