Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paip2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001902.1 | NP_001001902.1 | 3 - 4 | 497 | 96 | |
GenBank mRNA | BC068990.1 | - | 3 - 4 | 497 | 96 | |
CT030575.1 | - | 3 - 4 | 410 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |