Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004974.1 | NP_001004974.1 | 4 - 5 | 496 | 77 | |
GenBank mRNA | BC075495.1 | - | 4 - 5 | 496 | 77 | |
CR848421.2 | - | 4 - 5 | 532 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |