Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005080.1 | NP_001005080.1 | 2 - 3 | 405 | 95 | |
GenBank mRNA | BC077002.1 | - | 2 - 3 | 405 | 95 | |
CR761339.2 | - | 2 - 3 | 270 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |