Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp5pd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011021.2 | NP_001011021.1 | 4 - 5 | 366 | 78 | |
GenBank mRNA | BC082736.1 | - | 4 - 5 | 367 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |