Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011036.1 | NP_001011036.1 | 3 - 4 | 333 | 101 | |
XM_012960614.2 | XP_012816068.1 | 3 - 4 | 360 | 101 | ||
XM_012960619.2 | XP_012816073.1 | 3 - 4 | 360 | 101 | ||
GenBank mRNA | BC084062.1 | - | 3 - 4 | 333 | 101 | |
CR760777.2 | - | 3 - 4 | 323 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |