Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203623.1 | NP_988954.1 | 5 - 6 | 660 | 64 | |
XM_012970239.2 | XP_012825693.1 | 5 - 6 | 783 | 64 | ||
GenBank mRNA | BC061302.1 | - | 5 - 6 | 660 | 64 | |
BC089727.1 | - | 5 - 6 | 646 | 64 | ||
BC169133.1 | - | 643 | 64 | |||
CR760864.2 | - | 5 - 6 | 629 | 64 | ||
CR926445.2 | - | 5 - 6 | 459 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |