Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203719.1 | NP_989050.1 | 1 - 2 | 645 | 69 | |
GenBank mRNA | BC061344.1 | - | 1 - 2 | 645 | 69 | |
CR760510.2 | - | 1 - 2 | 420 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |