Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pin4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203870.1 | NP_989201.1 | 2 - 3 | 300 | 74 | |
XM_018096185.1 | XP_017951674.1 | 2 - 3 | 221 | 74 | ||
GenBank mRNA | BC063359.1 | - | 2 - 3 | 300 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |