Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204069.1 | NP_989400.1 | 2 - 3 | 202 | 65 | |
GenBank mRNA | AY336972.1 | - | 2 - 3 | 203 | 65 | |
BC064854.1 | - | 2 - 3 | 202 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |