Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005131.1 | NP_001005131.1 | 5 - 6 | 621 | 107 | |
XM_012960204.2 | XP_012815658.1 | 5 - 6 | 740 | 107 | ||
GenBank mRNA | BC077669.1 | - | 5 - 6 | 621 | 107 | |
CR760027.2 | - | 5 - 6 | 484 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |