Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrnp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005142.1 | NP_001005142.1 | 1 - 2 | 157 | 65 | |
GenBank mRNA | BC077690.1 | - | 1 - 2 | 157 | 65 | |
CT025161.2 | - | 1 - 2 | 55 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |