Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006691.1 | NP_001006692.1 | 4 - 5 | 672 | 75 | |
XM_012959488.2 | XP_012814942.1 | 5 - 6 | 780 | 75 | ||
XM_012959498.2 | XP_012814952.1 | 5 - 6 | 1386 | 75 | ||
XM_012959503.2 | XP_012814957.1 | 5 - 6 | 817 | 75 | ||
XM_012959507.2 | XP_012814961.1 | 6 - 7 | 1336 | 75 | ||
XM_012959516.2 | XP_012814970.1 | 5 - 6 | 715 | 75 | ||
GenBank mRNA | BC075392.1 | - | 4 - 5 | 672 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |