Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006812.1 | NP_001006813.1 | 125 | 63 | ||
XM_012967546.1 | XP_012823000.1 | 177 | 63 | |||
GenBank mRNA | BC076696.1 | - | 125 | 63 | ||
CR760722.2 | - | 102 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis |