Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdelr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008051.1 | NP_001008052.1 | 4 - 5 | 936 | 110 | |
GenBank mRNA | BC080938.1 | - | 4 - 5 | 936 | 110 | |
CR942706.2 | - | 4 - 5 | 638 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |