Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fhl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008164.1 | NP_001008165.1 | 2 - 3 | 285 | 90 | |
XM_012956824.2 | XP_012812278.1 | 3 - 4 | 435 | 90 | ||
XM_012956825.2 | XP_012812279.1 | 2 - 3 | 195 | 90 | ||
GenBank mRNA | BC082347.1 | - | 2 - 3 | 285 | 90 | |
CR760328.2 | - | 2 - 3 | 285 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |