Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011077.1 | NP_001011077.1 | 3 - 4 | 714 | 93 | |
XM_012960669.2 | XP_012816123.1 | 3 - 4 | 1204 | 93 | ||
GenBank mRNA | BC084462.1 | - | 3 - 4 | 714 | 93 | |
BC158260.1 | - | 3 - 4 | 717 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |