Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agpat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011265.1 | NP_001011265.1 | 678 | 147 | ||
GenBank mRNA | BC087973.1 | - | 678 | 147 |
Gene Symbol: LOC108647603 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018093995.1 | XP_017949484.1 | 1 - 2 | 194 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |