Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016201.2 | NP_001016201.1 | 101 | 73 | ||
GenBank mRNA | BC118697.1 | - | 1 - 2 | 115 | 73 | |
CR926344.2 | - | 101 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |