Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016609.1 | NP_001016609.1 | 3 - 4 | 303 | 82 | |
XM_012964710.2 | XP_012820164.1 | 4 - 5 | 800 | 82 | ||
XM_012964711.2 | XP_012820165.1 | 4 - 5 | 574 | 82 | ||
XM_012964712.2 | XP_012820166.1 | 4 - 5 | 800 | 82 | ||
GenBank mRNA | BC167412.1 | - | 4 - 5 | 364 | 82 | |
CR760531.1 | - | 3 - 4 | 303 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |