Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prelid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016922.2 | NP_001016922.1 | 1 - 2 | 111 | 101 | |
GenBank mRNA | BC091072.1 | - | 1 - 2 | 111 | 101 | |
CR848108.2 | - | 1 - 2 | 143 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |