Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prxl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017220.2 | NP_001017220.1 | 4 - 5 | 400 | 72 | |
XM_012966658.2 | XP_012822112.1 | 4 - 5 | 424 | 72 | ||
GenBank mRNA | CR760360.2 | - | 4 - 5 | 400 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Xenopus tropicalis Probe spans exons |